Anti-HP Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB97976-50EA 1100 GBP
ABCAAB97976-50
Anti-HP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HP (haptoglobin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR