Anti-Smad2 + Smad3 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB63672-100EA 1100 GBP
ABCAAB63672-100
Anti-Smad2 + Smad3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Smad2 + Smad3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR