Anti-MAML1 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB17019-100EA 1090 GBP
ABCAAB17019-100
Anti-MAML1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAML1 (mastermind-like 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR