Olive oil

Supplier: Merck

47118.EA 93.6 GBP
47118.
Olive oil
Olive oil
MDL Number: MFCD00131764
CAS Number: 8001-25-0
EINECS: 232-277-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR